Learn More
IMB-XH1, MedChemExpress
Product Code. 16471567
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471567 50 mg 50mg
16435378 10 mg 10mg
16418097 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16471567 Supplier Medchem Express Supplier No. HY12826
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
MedChemExpress IMB-XH1 is an inhibitor of myeloid cell factor 1 (Mcl-1). IMB-XH1 is a non-competitive Delhi metallo-β-lactamase (NDM-1) inhibitor. The IC50s of IMB-XH1 against metallo-β-lactamases NDM-1, IMP-4, ImiS and L1 are 0.4637 μM, 3.980 μM, 0.2287 μM and 1.158 μM, respectively.
IMB-XH1 is an inhibitor of myeloid cell factor 1 (Mcl-1). IMB-XH1 is a non-competitive Delhi metallo-β-lactamase (NDM-1) inhibitor. The IC50s of IMB-XH1 against metallo-β-lactamases NDM-1, IMP-4, ImiS and L1 are 0.4637 μM, 3.980 μM, 0.2287 μM and 1.158 μM, respectively.
Specifications
Chemical Name or Material IMB-XH1
CAS 292057-76-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H15N3OS
Quantity 50 mg
Solubility Information DMSO : 33.33 mg/mL (99.97 mM; Need ultrasonic)
SMILES OC1=C(N=CC=C2)C2=CC=C1C(NC3=CC=CC=N3)C4=CC=CS4
Molecular Weight (g/mol) 333.41
Formula Weight 333.41
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death