missing translation for 'onlineSavingsMsg'
Learn More

Imazalil, MedChemExpress

Product Code. 16428636
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16428636 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16428636 Supplier Medchem Express Supplier No. HYB1134
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Imazalil (Enilconazole) is a fungicide, widely used in agriculture, particularly in the growing of citrus fruits, also used in veterinary medicine as a topical antimycotic.

Imazalil (Enilconazole) is a fungicide, widely used in agriculture, particularly in the growing of citrus fruits, also used in veterinary medicine as a topical antimycotic.

Specifications

Chemical Name or Material Imazalil
Physical Form Solid
CAS 35554-44-0
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C14H14Cl2N2O
For Use With (Application) COVID-19-immunoregulation
Synonym Enilconazole
Solubility Information DMSO : ≥ 100 mg/mL (336.50 mM) ∣H2O : 20 mg/mL (67.30 mM; Need ultrasonic)
SMILES C=CCOC(C1=CC=C(Cl)C=C1Cl)CN2C=CN=C2
Molecular Weight (g/mol) 297.18
Formula Weight 297.18
Percent Purity 99.2%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.