Learn More
Cytiva TempliPhi™ 100/500 Amplification Kit
Product Code. 10794562 Shop All Cytiva Products
Change view
Click to view available options
No. of Reactions:
100
500
Unit Size:
1 set
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
10794562 100 1 set
10281390 500 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10794562 Supplier Cytiva Supplier No. 25640010
  • ONLINE EXCLUSIVE PRICE
    313.00 EUR
    249.80 EUR / 1 sets
    Save 63.20 EUR
    20% Off
 Restricted Item
In Stock
Includes
Sample buffer, Reaction buffer, Enzyme mix, and Positive control pUC19 DNA (2ng/μL)
Use amplified DNA directly for cycle sequencing without purification. Cytiva illustra TempliPhi DNA Amplification Kits efficiently prepare DNA sequencing templates from circular DNA.
  • Generate microgram quantities of template DNA from picogram amounts of starting material
  • Completed reactions contain uniform quantities of DNA irrespective of input amount, allowing for use directly in sequencing reactions without quantification
  • Amplify DNA from bacterial or M13 liquid cultures, colonies, plaques, glycerol stocks, or purified circular (plasmid or M13) DNA without the need for purification
  • Simple protocol reduces time, labor, and consumables needed for template preparation, and workflow enables easy automation
  • Contain separate, unmixed components for enhanced stability
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage The product can be stored for 2 months or less at -20°C. For storage longer than 2 months, store at -70°CThaw components on ice and maintain at 0–4°C during handling. DO NOT warm above 4°C prior to amplification
Description illustra TempliPhi DNA Amplification Kits efficiently prepare DNA sequencing templates from circular DNA.
Format Kit
For Use With (Application) Sequencing Template Amplification Kits and Reagents
Includes 0.5mL Sample Buffer, 0.5mL Reaction Buffer, 20μL Enzyme mix and 50μL positive control pUC19 DNA
No. of Reactions 100
Product Type illustra TempliPhi 100 Amplification Kit
Test Time 4-18 h depending on starting material