Learn More
Cytiva PuReTaq Ready-To-Go™ PCR Beads
Shop All Cytiva Products
Click to view available options
Format:
0.2mL Tube with hinged cap
0.2mL Tubes/plate
0.5mL Tube
No. of Reactions:
96
100
480
Volume:
200μL
500μL
Unit Size:
100 reactions
480 reactions
Each
Description
- puReTaq DNA polymerase and high-purity reagents provide robust performance with very low levels of contaminating DNA
- Manufacturing process locks in performance for excellent reproducibility with less risk of pipetting errors and contamination
- Long-term ambient-temperature stability simplifies shipping and removes any need for freezer storage
- Verified for real-time PCR
- Ready-To-Go means simply add template DNA and primers and cycle
Specifications
Specifications
| Content And Storage | Room temperature stable beads containing ∼ 2.5 units of recombinant PuReTaq DNA Polymerase, dATP, dCTP, dGTP, dTTP, stabilizers, BSA and reaction Buffer, When a bead is reconstituted to a 25μL final volume, the concentration of each dNTP |
| Format | 0.5mL Tube |
| Quantity | 1 Kit |
| For Use With (Application) | Standard PCR |
| No. of Reactions | 100 |
| Volume | 500μL |
Safety and Handling
Product Identifier
- PuReTaq
- EUH210-Safety data sheet available on request.
- UNKNOWN ACUTE-73.3 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 73.3% of components with unknown hazards to the aquatic environment.