missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ IL-2R ELISA Kit, Human

Product Code. 15388194
Change view
Click to view available options
Quantity:
192 Assays
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15388194 192 Assays Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15388194 Supplier Thermo Scientific™ Supplier No. EH2IL2R2

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

IL-2R ELISA Kit, Human

The Thermo Scientific Human IL-2R ELISA Kit is used to measure s-IL-2R (human soluble interleukin-2 receptor) levels in human serum, plasma (EDTA, heparin, sodium citrate) and culture supernatants. Each kit contains two anti-Human sIL-2R capture antibody precoated 96-well Strip Plates and sufficient detection antibody, conjugate, buffers, diluent, TMB and stop buffers, as well as lyophilized sIL-2R standard to analyze several samples.

Features of the Thermo Scientific Human IL-2R ELISA Kit:

  • Target: natural and recombinant human soluble interleukin-2 receptor (sIL-2R); also called CD25
  • Format: colorimetric ELISA (TMB substrate) in clear 96-well strip plate
  • Measurement: absorbance at 450nm minus absorbance at 550nm
  • Assay Range: 0 to 6500 U/mL
  • Sensitivity: < 24 U/mL
  • Precision: intra-assay CV ≤ 10%; inter-assay CV ≤ 10%
  • Sample Types: cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
  • Total Assay Time: 2.5 hours
  • Sample Size: 50μL
  • Specificity: no significant cross-reaction with human IL-1α, IL-1β, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-10, IL-11, IL-12, IL-13, IL-15, GM-CSF, IFNγ, IFNα, TNFβ, or mouse IL-2.

    Note: Standards in this ELISA are calibrated to an internal sIL-2R reference preparation of supernatant from a transfected cell line that secretes recombinant soluble IL-2R. We define 1000 units as the amount of sIL-2R present in 1 mL of an internal reference preparation. Units/mL can be converted to sIL-2R pg/mL using the following formula: (sIL-2R pg/mL) = (sIL-2R U/mL) x (1/0.113).

    This human IL-2 receptor ELISA kit uses pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of soluble interleukin-2 receptor (sIL-2R, also called CD25) in samples at 50μL per well. Also included are calibrated recombinant protein standard, TMB substrate solution and stop solution. The complete kit contains pre-coated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.

    Includes:
    Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Assay Range 0 to 6500U/mL
    Assay Sensitivity <24U/mL
    Description IL-2R ELISA Kit, 192 Assays, Human Species, 0 to 6500 U/mLA€  Assay Range, Cytokines and Receptors Protein Family, Colorimetric Detection Method, For Protein Assays and Analysis
    Detection Method Colorimetric
    Immunoassay Kit Format Strip Plate
    Product Type IL-2R ELISA Kit
    Protein Family Cytokines and receptors
    Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
    Specificity No significant cross-reaction with human IL-1α±, IL-1β, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-10, IL-11, IL-12, IL-13, IL-15, GM-CSF, IFNγ, IFNα±, TNFβ, or mouse IL-2
    Target Species Human
    For Use With (Application) Protein assays and analysis
    Kit Contents Detector antibody, Diluents, HRP Conjugate, Pre-coated plate(s), Standard or calibrator, Stop solution, Substrate, Wash buffer
    Measurement Absorbance at 450nm minus absorbance at 550nm
    Precision Intra-assay CV ≤10%; inter-assay CV ≤10%
    Quantity 192 Assays
    Regulatory Status For research use only (RUO)
    Research Discipline Immunology
    Sample Volume 50 μL
    Storage Requirements 2°C to 8°C
    Target CD25
    Total Assay Time 2.5 hr.
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.