missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IkB-beta Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
0.5 mg
100μg
Unit Size:
0.5mg
100µg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-356 of Human IkB-beta The Recombinant Human IkB-beta Protein is derived from E. coli. The Recombinant Human IkB-beta Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Concentration | 1 mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | Liquid. 20 mM Tris-HCl buffer (pH 8.0) containing 20% glycerol, 1 mM DTT |
| Molecular Weight (g/mol) | 40.3 kDa |
| Purification Method | SDS-PAGE |
| Quantity | 0.5 mg |
| Source | Human |
| Storage Requirements | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Regulatory Status | RUO |
| Common Name | IkB-beta |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction