missing translation for 'onlineSavingsMsg'
Learn More

IkB-alpha, p Ser32 Antibody (SR1656), Novus Biologicals™

Rabbit Monoclonal Antibody

338.00 EUR - 449.00 EUR

Specifications

Antigen IkB-alpha
Clone SR1656
Dilution Western Blot 1:500-1:2000, Immunoprecipitation 1:50
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18663995
View Documents
Novus Biologicals
NBP3-21533-100ul
100 μg
449.00 EUR
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18602314
View Documents Promotion Details
Novus Biologicals
NBP3-21533-50ul
50 μg
338.00 EUR
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IkB-alpha Monoclonal antibody specifically detects IkB-alpha in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications

Specifications

IkB-alpha
Western Blot 1:500-1:2000, Immunoprecipitation 1:50
Monoclonal
Purified
RUO
Human
alpha, I-kappa-B-alpha, IkappaBalphaNF-kappa-B inhibitor alpha, IKBAikB-alpha, IkB-alpha, MAD3, MAD-3, Major histocompatibility complex enhancer-binding protein MAD3, NFKBI, nuclear factor of kappa light chain gene enhancer in B-cells, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor
A synthesized peptide derived from human phospho-IKB alpha [Ser32] (Uniprot #: P25963)
Primary
Store at -20 degrees C. Avoid freeze/thaw cycles.
SR1656
Western Blot, Immunoprecipitation
Unconjugated
Rabbit
Apoptosis, Cell Biology, Chaperone Mediated Autophagy (CMA), Diabetes Research, DNA Repair, Nucleotide Excision Repair, Phospho Specific, Signal Transduction
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
4792
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.