missing translation for 'onlineSavingsMsg'
Learn More

IkB-alpha, p Ser32 Antibody (SR1656), Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-21533-50ul

289.00 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.


 View more versions of this product

Product Code. 18602314

  • 338.00 EUR / 50µL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

IkB-alpha Monoclonal antibody specifically detects IkB-alpha in Human samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IkB-alpha
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot, Immunoprecipitation
SR1656
Western Blot 1:500-1:2000, Immunoprecipitation 1:50
alpha, I-kappa-B-alpha, IkappaBalphaNF-kappa-B inhibitor alpha, IKBAikB-alpha, IkB-alpha, MAD3, MAD-3, Major histocompatibility complex enhancer-binding protein MAD3, NFKBI, nuclear factor of kappa light chain gene enhancer in B-cells, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor
A synthesized peptide derived from human phospho-IKB alpha [Ser32] (Uniprot #: P25963)
50 μg
Apoptosis, Cell Biology, Chaperone Mediated Autophagy (CMA), Diabetes Research, DNA Repair, Nucleotide Excision Repair, Phospho Specific, Signal Transduction
4792
Store at -20 degrees C. Avoid freeze/thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.