Description
- Clamping range, stand: 34mm
- Clamping range, extension arm: 16mm
- Manufactured from cast aluminum
Specifications
Specifications
| Diameter (English) | 0.6 in. |
| Diameter (Metric) | 16 mm |
| For Use With (Equipment) | For the Stands R 2722 and R 2723 as Well as Extensions |
| Type | Boss Head Clamp |