Learn More
Proteintech IHCeasy NFATC4 Ready-To-Use IHC Kit
Product Code. 18045595 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
50 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18045595 50 Tests 50 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18045595 Supplier Proteintech Supplier No. KHC174050T
KHC1740 is a ready-to-use IHC kit for staining of NFATC4.
KHC1740 is a ready-to-use IHC kit for staining of NFATC4. The kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use. Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Specifications
Quantity 50 Tests
Form Liquid