Description
KHC1740 is a ready-to-use IHC kit for staining of NFATC4. The kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use. Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Specifications
Specifications
| Quantity | 50 Tests |
| Form | Liquid |