missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGF2BP3 Monoclonal Antibody (OTI6A4), TrueMAB™, OriGene
Description
IGF2BP3 Monoclonal antibody specifically detects IGF2BP3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
| Antigen | IGF2BP3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI6A4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | IGF2BP3 |
| Gene Accession No. | O00425 |
| Gene Alias | 2610101N11Rik; AA522010; Ab2-255; AL022933; AU045931; cancer/testis antigen 98; CT98; hKOC; IGF2 mRNA-binding protein 3; IGF2BP3; IGF-II mRNA-binding protein 3; IMP3; IMP-3; insulin like growth factor 2 mRNA binding protein 3; insulin-l; insulin-like growth factor 2 mRNA binding protein 3; insulin-like growth factor 2 mRNA-binding protein 3; insulin-like growth factor 2, binding protein 3; KH domain containing protein overexpressed in cancer; KH domain-containing protein overexpressed in cancer; KOC; KOC1; Koc13; mimp3; mIMP-3; Neilsen; RGD1306512; VICKZ family member 3; Vickz3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?