Learn More
Brady™ IDXPERT™ B-427 Self-Laminating Voice and Data Communication Cable Marker Labels
Product Code. 11594344 Shop All Brady Products
Change view
Click to view available options
Color:
Black on White
Black on Yellow
Dimensions (L x W):
0.5 x 0.75 in. (W x H)
0.5 x 1 in. (W x H)
0.5 x 1.25 in. (W x H)
0.5 x 1.5 in. (W x H)
0.75 x 1 in. (W x H)
0.75 x 1.5 in. (W x H)
1 x 1 in.
1 x 1.25 in. (W x H)
1 x 1.5 in. (W x H)
1 x 2.5 in. (W x H)
1 x 4 in. (W x H)
1.5 x 1.25 in. (W x H)
1.5 x 1.5 in.
1.5 x 2.5 in. (W x H)
1.5 x 4 in. (W x H)
1.5 x 6 in. (W x H)
Packungsgröße:
1 roll
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Color Dimensions (L x W) Packungsgröße
11594344 Black on White 0.5 x 1 in. (W x H) 1 roll
11584344 Black on White 0.5 x 1.25 in. (W x H) 1 roll
11524344 Black on White 1 x 1.25 in. (W x H) 1 roll
11504354 Black on White 0.5 x 0.75 in. (W x H) 1 roll
11594334 Black on White 1.5 x 1.25 in. (W x H) 1 roll
11554334 Black on White 1.5 x 2.5 in. (W x H) 1 roll
11564344 Black on White 0.75 x 1 in. (W x H) 1 roll
10561845 Black on Yellow 1 x 1 in. 1 roll
11564334 Black on White 1 x 2.5 in. (W x H) 1 roll
11574334 Black on Yellow 1 x 2.5 in. (W x H) 1 roll
11534334 Black on White 1 x 4 in. (W x H) 1 roll
11544334 Black on Yellow 1 x 4 in. (W x H) 1 roll
11574344 Black on Yellow 0.5 x 1.5 in. (W x H) 1 roll
11504344 Black on White 1 x 1.5 in. (W x H) 1 roll
11534344 Black on Yellow 0.75 x 1.5 in. (W x H) 1 roll
11514344 Black on Yellow 1 x 1.5 in. (W x H) 1 roll
11584334 Black on White 1.5 x 1.5 in. 1 roll
11524334 Black on White 1.5 x 4 in. (W x H) 1 roll
11514334 Black on White 1.5 x 6 in. (W x H) 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11594344 Lieferant Brady™ Lieferanten-Nr. 800768
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
For use with IDEXPERT printers. Brady Self-Laminating Vinyl Labels are used for wire and cable marking, panel identification and voice/data identification
  • Printable, self-laminating vinyl labels for the IDXPERT printer
  • Material is UL recognized
  • Used for mounting to rigid panels
Spezifikation
Product Type IDXPERT Label
Color Black on White
For Use With (Equipment) IDXPERT Label Printer
Material Self-laminating Vinyl (B-427)
Width (Metric) 12.70 mm
Height (Metric) 25.40 mm
Thickness (English) 0.004 in.
Thickness (Metric) 0.102 mm
Dimensions (L x W) 0.5 x 1 in. (W x H)
Adhesive Type Permanent
Description Conformability, Good Clarity, Self-Extinguishing, Self-Laminating material
Finish Matte
Shape Rectangle
Temperature Range (Metric) -70°C to +70°C
Certifications/Compliance UL Recognized
Quantity 450/Roll
Mehr anzeigen Weniger anzeigen