missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDH2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
226.00 EUR - 484.00 EUR
Specifications
| Antigen | IDH2 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1 - 2 ug/mL, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence 1:250 - 1:500, Immunohistochemistry-Paraffin 1:100 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18129171
|
Novus Biologicals
NBP2-22166SS |
0.025 mL |
226.00 EUR
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18138711
|
Novus Biologicals
NBP2-22166 |
0.1 mL |
484.00 EUR
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IDH2 Polyclonal specifically detects IDH2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| IDH2 | |
| Western Blot 1 - 2 ug/mL, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence 1:250 - 1:500, Immunohistochemistry-Paraffin 1:100 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Epigenetics, Hypoxia, Lipid and Metabolism | |
| PBS with 0.02% Sodium Azide | |
| D2HGA2, EC 1.1.1.42, ICD-M, IDH, IDHM, IDP, IDPM, isocitrate dehydrogenase [NADP], mitochondrial, isocitrate dehydrogenase 2 (NADP+), mitochondrial, mNADP-IDH, NADP(+)-specific ICDH, Oxalosuccinate decarboxylase | |
| IDH2 | |
| IgG | |
| Affinity Purified | |
| Human, mouse and rat. Predicted to react with monkey, hamster, Guinea pig and rabbit based on 100% sequence homology. |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| P48735, P54071, P56574 | |
| 3418 | |
| A synthetic peptide made to a C-terminal portion of the human IDH2 protein (between residues 375-452) [UniProt P48735]. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title