missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDH2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NBP2-22166
This item is not returnable.
View return policy
Description
IDH2 Polyclonal specifically detects IDH2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| IDH2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| D2HGA2, EC 1.1.1.42, ICD-M, IDH, IDHM, IDP, IDPM, isocitrate dehydrogenase [NADP], mitochondrial, isocitrate dehydrogenase 2 (NADP+), mitochondrial, mNADP-IDH, NADP(+)-specific ICDH, Oxalosuccinate decarboxylase | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, mouse and rat. Predicted to react with monkey, hamster, Guinea pig and rabbit based on 100% sequence homology. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1 - 2 ug/mL, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence 1:250 - 1:500, Immunohistochemistry-Paraffin 1:100 - 1:500 | |
| P48735, P54071, P56574 | |
| IDH2 | |
| A synthetic peptide made to a C-terminal portion of the human IDH2 protein (between residues 375-452) [UniProt P48735]. | |
| 0.1 mL | |
| Epigenetics, Hypoxia, Lipid and Metabolism | |
| 3418 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction