missing translation for 'onlineSavingsMsg'
Learn More

IDH-305, MedChemExpress

Product Code. 16484956
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
1mg
200mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16484956 100 mg 100mg
16433938 10 mg 10mg
16430986 200 mg 200mg
16416657 5 mg 5mg
16410297 50 mg 50mg
16401049 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16484956 Supplier Medchem Express Supplier No. HY104036

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress IDH-305 is an orally available, mutant-selective and brain-penetrant IDH1 inhibitor that targets IDH1 (R132) mutation. IDH-305 exhibits greater than 200 fold selectivity for mutant IDH1 isoforms vs. WT (IC50= 27 nM (IDH1R132H), 28 nM (IDH1R132C), 6.14 μM (IDH1WT)).

IDH-305 is an orally available, mutant-selective and brain-penetrant IDH1 inhibitor that targets IDH1 (R132) mutation. IDH-305 exhibits greater than 200 fold selectivity for mutant IDH1 isoforms vs. WT (IC50= 27 nM (IDH1R132H), 28 nM (IDH1R132C), 6.14 μM (IDH1WT)).

Specifications

Chemical Name or Material IDH-305
CAS 1628805-46-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H22F4N6O2
Quantity 100 mg
Solubility Information DMSO : ≥ 150 mg/mL (305.84 mM)
SMILES O=C1OC[C@H]([C@@H](F)C)N1C2=NC(N[C@H](C3=CC(C)=C(C4=CC(C(F)(F)F)=NC=C4)C=N3)C)=NC=C2
Molecular Weight (g/mol) 490.45
Formula Weight 490.45
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.