Learn More
ICS Analytes Sub-B1: Be, Co, Cu, Mn @ 50; Ag, Cd, Ni, Pb, Zn @ 100 μg/mL in 5% HNO3, VHG™
Description
ICS Analytes Sub-B1: Be, Co, Cu, Mn @ 50; Ag, Cd, Ni, Pb, Zn @ 100 μg/mL in 5% HNO3
Specifications
Specifications
| Chemical Name or Material | Beryllium/Cobalt/Copper/Manganese/Cadmium/Lead/Nickel/Silver/Zinc |
| CAS | 7440-41-7, 7440-48-4, 7440-50-8, 7439-96-5, 7440-43-9, 7439-92-1 |
| Methods | ICP |
| Analyte or Component Names | Beryllium/Cobalt/Copper/Manganese/Cadmium/Lead/Nickel/Silver/Zinc |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute HNO3 |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Beryllium 50 μg/mL/Cobalt 50 μg/mL/Copper 50 μg/mL/Manganese 50 μg/mL/Cadmium 100 μg/mL/Lead 100 μg/mL/Nickel 100 μg/mL/Silver 100 μg/mL/Zinc 100 μg/mL |