Learn More
Icaridin (Picaridin), Mikromol™
Product Code. 17923216
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17923216 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17923216 Supplier Mikromol Supplier No. MM3755.00
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Icaridin (Picaridin)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Picaridin
CAS 119515-38-7
Analyte or Component Names Picaridin
Shipping Condition Room Temperature
Molecular Formula C12 H23 N O3
Certifications/Compliance ISO 17034
Synonym 2-(2-Hydroxyethyl)-1-piperidinecarboxylic Acid 1-Methylpropyl Ester, 1-Methylpropyl 2-(2-hydroxyethyl)-1-piperidinecarboxylate, Autan, Autan Repel, Bayrepel, Icaridin, Isobutyl 2-(2-hydroxyethyl)-1-piperidinecarboxylate, KBR 3023, Natrapel, Picaridin, Pikaridin, Propidine
InChI Formula InChI=1S/C12H23NO3/c1-3-10(2)16-12(15)13-8-5-4-6-11(13)7-9-14/h10-11,14H,3-9H2,1-2H3
SMILES CCC(C)OC(=O)N1CCCCC1CCO
IUPAC Name butan-2-yl 2-(2-hydroxyethyl)piperidine-1-carboxylate
Molecular Weight (g/mol) 229.32
Quantity 250 mg
Formula Weight 229.1678 g/mol