missing translation for 'onlineSavingsMsg'
Learn More

Ibufenac Acyl-Beta-D-Glucuronide, TRC

Product Code. 30180382
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180382 1 mg 1mg
30180004 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30180382 Supplier Toronto Research Chemicals Supplier No. TRCI1300051MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 164669-89-0
Molecular Formula C18H24O8
Molecular Weight (g/mol) 368.38
Synonym β-D-Glucopyranuronic acid 1-[4-(2-methylpropyl)benzeneacetate]
SMILES O=C(CC1=CC=C(CC(C)C)C=C1)O[C@H]([C@@H]2O)OC(C(O)=O)[C@@H](O)[C@@H]2O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C18H24O8/c1-9(2)7-10-3-5-11(6-4-10)8-12(19)25-18-15(22)13(20)14(21)16(26-18)17(23)24/h3-6,9,13-16,18,20-22H,7-8H2,1-2H3,(H,23,24)/t13-,14-,15+,16?,18-/m0/s1
Percent Purity >95%
Chemical Name or Material Ibufenac Acyl-beta-D-Glucuronide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.