CAS RN 164669-89-0
CAS RN 164669-89-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ibufenac Acyl-Beta-D-Glucuronide, TRC
CAS: 164669-89-0 Molecular Formula: C18H24O8 Molecular Weight (g/mol): 368.38 Synonym: β-D-Glucopyranuronic acid 1-[4-(2-methylpropyl)benzeneacetate] SMILES: O=C(CC1=CC=C(CC(C)C)C=C1)O[C@H]([C@@H]2O)OC(C(O)=O)[C@@H](O)[C@@H]2O