missing translation for 'onlineSavingsMsg'
Learn More

Ibrutinib, MedChemExpress

Product Code. 16474338
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
200mg
500mg
50mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16474338 10 mg 10mg
16445256 100 mg 100mg
16430667 50 mg 50mg
16421596 500 mg 500mg
16421016 200 mg 200mg
16431139 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16474338 Supplier Medchem Express Supplier No. HY10997
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Ibrutinib (PCI-32765) is a selective, irreversible Btk inhibitor with an IC50 of 0.5 nM.

Ibrutinib (PCI-32765) is a selective, irreversible Btk inhibitor with an IC50 of 0.5 nM.

Specifications

Chemical Name or Material Ibrutinib
CAS 936563-96-1
Color White
Purity Grade Notes Research
Molecular Formula C25H24N6O2
Quantity 10 mg
Synonym PCI-32765
Solubility Information DMSO : 100 mg/mL (227.01 mM; Need ultrasonic)
SMILES C=CC(N1C[C@H](N2N=C(C3=CC=C(OC4=CC=CC=C4)C=C3)C5=C(N)N=CN=C52)CCC1)=O
Molecular Weight (g/mol) 440.5
Formula Weight 440.5
Percent Purity 98.07%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.