Learn More
Brady™ i3300 Industrial Label Printer EU with Wi-Fi
Product Code. 16306925 Shop All Brady Products
Change view
Click to view available options
BradyPrinter i3300 EU with Wi-Fi; Print Speed: 101.6 mm/s; Resolution: 300 dpi; Print Technology: Thermal Transfer; Weight (Metric): 5.2 kg; Color Capability: Monochrome
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16306925 BradyPrinter i3300 EU with Wi-Fi; Print Speed: 101.6 mm/s; Resolution: 300 dpi; Print Technology: Thermal Transfer; Weight (Metric): 5.2 kg; Color Capability: Monochrome
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16306925 Supplier Brady™ Supplier No. 149554
Estimated Shipment: 04-12-2026
The BradyPrinter i3300 Industrial Label Printer can print a wide range of sign and labels in monocolor, including labels to identify wires, products, laboratory samples and facilities.
This mid-volume industrial label printer is ideal for identification throughout your facility, including everything from wires and panels to safety and facility labels.
  • Wi-Fi connectivity allows you to get data more easily from a variety of sources
  • Print smart technology allows the printer to print on the first label, even on sleeves and small labels
  • Foolproof drop-in label rolls and ribbons snap into place in 20 seconds
  • Prints on more than 825 parts and 70 material options for facility-wide identification
  • Contact our Brady™ Technical Support Team to schedule a free phone appointment
  • Comes with QWERTY keyboard
Specifications
Display Type Color LCD Touchscreen
Print Technology Thermal Transfer
Width (English) 9 in.
Width (Metric) 229 mm
Height (English) 9.5 in.
Height (Metric) 241 mm
Depth (English) 12 in.
Depth (Metric) 305 mm
Autoclavable Not Autoclavable
Color Capability Monochrome
Includes i3300 Printer, EU Power Cord, Brady™ Workstation Basic Suite, USB Cable, Product CD, Cleaning Kit, Printed Quick Start Guide
Languages Bulgarian, Chinese (Traditional), Chinese (Simplified), Croatian, Czech, Danish, Dutch, English, Estonian, Finnish, French, German, Hungarian, Italian, Norwegian, Polish, Portuguese, Romanian, Russian, Spanish, Swedish, Turkish
Memory 195 MB
Model i3300
Print Speed 101.6 mm/s
Resolution 300 dpi
Connector Types Ethernet, USB, USB 2.0
Operating System Windows™ 10, Windows™ 7, Windows™ 8, Windows™ Servers 2012, 2012 R2, and 2016
Temperature Range (Metric) 10°C to 40°C
Weight (English) 11.5 lb.
Weight (Metric) 5.2 kg
For Use With (Application) Circuit Board and Barcode Labeling, Component Labeling, General Identification
Warranty 1 Year
Voltage 110/240 V
Alternate Part Number i3300-C-EU-WF