Learn More
- Home
- Products
- Chromatography Columns and Cartridges
- HPLC Columns and Cartridges
- HPLC Columns
- Thermo Scientific™ HyPURITY™ C18 HPLC Columns, 3μm Particle Size
Thermo Scientific™ HyPURITY™ C18 HPLC Columns, 3μm Particle Size
Product Code. p-4522847 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
10 mm
30 mm
50 mm
100 mm
150 mm
250 mm
Diameter (Metric):
2.1 mm
3 mm
4 mm
4.6 mm
Particle Size:
3 μm
5 μm
Unit Size:
Each
Pack of 4
27 product options available for selection
| Product Code. | Length (Metric) | Diameter (Metric) | Particle Size | Unit Size |
|---|---|---|---|---|
| 10096084 | 50 mm | 2.1 mm | 3 μm | Each |
| 10648445 | 50 mm | 4.6 mm | 3 μm | Each |
| 10109194 | 100 mm | 2.1 mm | 3 μm | Each |
| 10483285 | 100 mm | 3 mm | 3 μm | Each |
| 10290755 | 100 mm | 4.6 mm | 3 μm | Each |
| 10648615 | 150 mm | 2.1 mm | 3 μm | Each |
| 10118954 | 150 mm | 3 mm | 3 μm | Each |
| 10392965 | 150 mm | 4.6 mm | 3 μm | Each |
| 10704437 | 10 mm | 2.1 mm | 3 μm | Pack of 4 |
| 10006114 | 10 mm | 4 mm | 3 μm | Pack of 4 |
| 10690236 | 10 mm | 2.1 mm | 5 μm | Pack of 4 |
| 10751937 | 10 mm | 3 mm | 5 μm | Pack of 4 |
| 10761937 | 10 mm | 4 mm | 5 μm | Pack of 4 |
| 10017514 | 30 mm | 2.1 mm | 5 μm | Each |
| 10477634 | 50 mm | 2.1 mm | 5 μm | Each |
| 10230615 | 50 mm | 3 mm | 5 μm | Each |
| 10077554 | 50 mm | 4.6 mm | 5 μm | Each |
| 10231195 | 100 mm | 2.1 mm | 5 μm | Each |
| 10270905 | 100 mm | 4 mm | 5 μm | Each |
| 10408384 | 100 mm | 4.6 mm | 5 μm | Each |
| 10601116 | 150 mm | 2.1 mm | 5 μm | Each |
| 10067644 | 150 mm | 3 mm | 5 μm | Each |
| 10399243 | 150 mm | 4 mm | 5 μm | Each |
| 10630346 | 150 mm | 4.6 mm | 5 μm | Each |
| 10518795 | 250 mm | 2.1 mm | 5 μm | Each |
| 10077454 | 250 mm | 4 mm | 5 μm | Each |
| 10528795 | 250 mm | 4.6 mm | 5 μm | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 27 options available.
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Codice prodotto.
10096084
Fornitore
Thermo Scientific™
N. del fornitore
22103052130
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
Thermo ScientificTM HyPURITYTM C18 HPLC Columns are available in limited dimensions. Please contact Customer Service for more details.
- Highly characterized, tested and validated C18 columns
Ordering Information:
Other column dimensions are also available. Please call Customer Service for more information.
Specifica
Specifica
| Column Format | Analytical Column |
| Diameter (Metric) | 2.1 mm |
| For Use With | LC/MS |
| Length (Metric) | 50 mm |
| Particle Size | 3 μm |
| pH | 1 to 9 |
| Pore Size | 190 Å |
| Surface Area | 200 m²/g |
| USP Type | L1 |
| Stationary Phase | C18 |
| Packing Material | Silica |
| Carbon Load | 13% |
| Column Type | Reversed Phase |
| Endcapped | Yes |
| Phase | Reversed Phase |
| Vedi altri risultati Mostra meno risultati |
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?