Learn More
Digi-Sense Hypodermic Thermocouple, Grounded d, Type T
Product Code. 11755288 Shop All Cole Parmer Products
Change view
Click to view available options
Analysis Time:
0.5 to 1.2 sec. or greater
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Analysis Time Unit Size
11755288 0.5 to 1.2 sec. or greater Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11755288 Supplier Digi-Sense Supplier No. 0811365
  • ONLINE EXCLUSIVE PRICE
    129.00 EUR
    122.50 EUR / Pack of 1
    Save 6.50 EUR
    5% Off
Estimated Shipment: 16-02-2027
This Hypodermic Thermocouple is a hypodermic tip penetration probe designed for soft and semi-soft materials.
  • Ideal for checking food temperatures.
  • Response Time: 0.5 to 1.2 seconds or greater.
  • Probe comes with Stainles Steel Handle.
  • Each probe comes with color coded ANSI miniconnectors: type J-black, type K-yellow, and type T-blue.
  • Note: the temperature range is for the probe tip only.
  • The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material.
  • ABS handles are rated to 220°F (104°C).
  • PVC handles are rated to 200°F (93°C).
  • Stainless Steel handles are rated at 450°F (232°C).
  • Mini Connectors are rated to 392°F (200°C).
  • Custom designs or changes to this product are available upon request.
  • Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Specifications
Analysis Time 0.5 to 1.2 sec. or greater
Certifications/Compliance ANSI
Temperature (Metric) -200°C to +232°C
Type Thermocouple Probe
For Use With (Application) Checking Food Temperatures
Style Type T
Temperature (English) -328°F to +449.6°F
Length (English) 4 in.
Length (Metric) 10.16 cm
Diameter (Metric) Process Tube 1.6 mm