Learn More
uvex Hypervision CA FBL. AF GY/GY/BU
Product Code. 31751170 Shop All Uvex Products
Change view
Click to view available options
Lens Material:
Polycarbonate
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lens Material Unit Size
31751170 Polycarbonate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31751170 Supplier uvex Supplier No. 9321614
  • ONLINE EXCLUSIVE PRICE
    16.80 EUR
    16.38 EUR / Pack of 1
    Save 0.42 EUR
    2% Off
Estimated Shipment: 26-11-2026
Goggles uvex hypervision Clear 9321614
  • The uvex hypervision features a comfortable, ergonomic design with a translucent frame that provides an almost unrestricted field of vision for enhanced workplace awareness.
  • A soft, flexible frame construction ensures high wearer comfort, even during extended periods of use.
  • Integrated side recesses allow sufficient space for personal prescription spectacles, enabling the uvex hypervision to be worn comfortably as reliable overspecs.
  • An adjustable headband combined with indirect ventilation completes the design, delivering a secure fit, improved airflow, and dependable eye protection for everyday professional applications.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant, Impact-resistant, Anti-fog
Lens Tint None
Product Type Goggles
Material Plastic
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection None
Frame Type Frameless