Learn More
Thermo Scientific™ Hypersil™ Silica HPLC Columns
Product Code. 10273593 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
10 mm
50 mm
100 mm
125 mm
150 mm
200 mm
250 mm
Diameter (Metric):
2.1 mm
3 mm
4 mm
4.6 mm
Particle Size:
3 μm
5 μm
Unit Size:
Each
Pack of 4
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Particle Size Unit Size
10273593 150 mm 4.6 mm 3 μm Each
10417073 50 mm 4.6 mm 5 μm Each
11533081 125 mm 4 mm 3 μm Each
10263593 150 mm 2.1 mm 3 μm Each
10119334 150 mm 4 mm 5 μm Each
10213313 150 mm 4.6 mm 5 μm Each
10713026 250 mm 4 mm 5 μm Each
10725973 250 mm 4.6 mm 5 μm Each
10334223 50 mm 4.6 mm 3 μm Each
10221295 100 mm 2.1 mm 5 μm Each
11587431 200 mm 4.6 mm 5 μm Each
10354523 50 mm 2.1 mm 3 μm Each
10180843 10 mm 2.1 mm 3 μm Pack of 4
10544665 10 mm 3 mm 3 μm Pack of 4
10098762 10 mm 4 mm 3 μm Pack of 4
10416883 10 mm 4 mm 5 μm Pack of 4
10350865 10 mm 3 mm 5 μm Pack of 4
10588405 100 mm 4.6 mm 3 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Product Code. 10273593 Supplier Thermo Scientific™ Supplier No. 30003154630
Estimated Shipment: 12-11-2026
Use the efficient and selective Thermo Scientific™ Hypersil™ Silica LC Columns for normal phase chromatography of nonpolar and moderately polar organic compounds.
Use the efficient and selective Thermo Scientific™ Hypersil™ Silica LC Columns for normal phase chromatography of nonpolar and moderately polar organic compounds.
Hypersil stationary phases are recognized as an industry standard in HPLC. Proven as an effective analytical tool, classical Hypersil phases are well established and referenced in HPLC methods. Quality is assured by manufacture under ISO 9001-2000 quality guidelines.
For more challenging separations that require improved chromatographic performance, use Thermo Scientific™ Hypersil GOLD™ Silica LC Columns as an alternative.
  • Excellent batch-to-batch reproducibility
  • Spherical particle with narrow particle size distribution
  • Long column lifetime
  • High performance and column efficiency
Ordering Information:
Other column dimensions are available. Also available in 10μm. Please call your Customer Service Representative for more information.
TRUSTED_SUSTAINABILITY
Specifications
Column Format Analytical Column
Diameter (Metric) 4.6 mm
For Use With HPLC
Length (Metric) 150 mm
Max. Pressure 5800 psi (400 bar)
Particle Size 3 μm
pH 2 to 8
Pore Size 120 Å
Surface Area 170 m²/g
Temperature 60°C
USP Type L3
Stationary Phase Silica
Product Line Hypersil Classical
Packing Material Spherical, Fully Porous Type A Silica
Column Type Normal Phase
Phase Normal Phase
Product Type Analytical Column