Learn More
- Home
- Products
- Chromatography Columns and Cartridges
- HPLC Columns and Cartridges
- HPLC Columns
- Thermo Scientific™ Hypersil™ BDS C18 Columns
Thermo Scientific™ Hypersil™ BDS C18 Columns
Product Code. 10662674 Shop All Thermo Scientific Products| Product Code. | Length (Metric) | Diameter (Metric) | Particle Size | Column Format | unitSize |
|---|---|---|---|---|---|
| 10662674 | 50 mm | 2.1 mm | 5 μm | Analytical Column | Each |
| 10670136 | 50 mm | 3 mm | 5 μm | Analytical Column | Each |
| 16308586 | 250 mm | 10 mm | 5 μm | Preparative Column, Standard | Each |
| 10079192 | 50 mm | 4.6 mm | 5 μm | Analytical Column | Each |
| 10067244 | 50 mm | 4 mm | 3 μm | Analytical Column | Each |
| 10203423 | 100 mm | 2.1 mm | 5 μm | Analytical Column | Each |
| 10488004 | 100 mm | 3 mm | 5 μm | Analytical Column | Each |
| 10610156 | 100 mm | 4 mm | 5 μm | Analytical Column | Each |
| 10110513 | 100 mm | 4.6 mm | 5 μm | Analytical Column | Each |
| 10389543 | 125 mm | 3 mm | 5 μm | Analytical Column | Each |
| 11593041 | 125 mm | 4.6 mm | 3 μm | Analytical Column | Each |
| 10097544 | 125 mm | 4 mm | 5 μm | Analytical Column | Each |
| 10217423 | 125 mm | 4.6 mm | 5 μm | Analytical Column | Each |
| 10484983 | 150 mm | 2.1 mm | 5 μm | Analytical Column | Each |
| 10616185 | 150 mm | 3 mm | 5 μm | Analytical Column | Each |
| 10403475 | 150 mm | 4 mm | 5 μm | Analytical Column | Each |
| 10283703 | 150 mm | 4.6 mm | 5 μm | Analytical Column | Each |
| 10438964 | 200 mm | 4.6 mm | 5 μm | Analytical Column | Each |
| 10503425 | 250 mm | 2.1 mm | 5 μm | Analytical Column | Each |
| 10494435 | 250 mm | 3 mm | 5 μm | Analytical Column | Each |
| 10468054 | 250 mm | 4 mm | 5 μm | Analytical Column | Each |
| 10344413 | 250 mm | 4.6 mm | 5 μm | Analytical Column | Each |
| 10734414 | 10 mm | 3 mm | 3 μm | Guard Cartridge, 4 Pk. | Pack of 4 |
| 10350885 | 150 mm | 4.6 mm | 2.4 μm | Analytical Column | Each |
| 10340885 | 150 mm | 2.1 mm | 2.4 μm | Analytical Column | Each |
| 10075964 | 100 mm | 4.6 mm | 2.4 μm | Analytical Column | Each |
| 10066694 | 100 mm | 2.1 mm | 2.4 μm | Analytical Column | Each |
| 10279864 | 50 mm | 4.6 mm | 2.4 μm | Analytical Column | Each |
| 10621416 | 50 mm | 2.1 mm | 2.4 μm | Analytical Column | Each |
| 10100943 | 10 mm | 4 mm | 5 μm | Guard Cartridge, 4 Pk. | Pack of 4 |
| 10764047 | 10 mm | 3 mm | 5 μm | Guard Cartridge, 4 Pk. | Pack of 4 |
| 10601144 | 10 mm | 2.1 mm | 5 μm | Guard Cartridge, 4 Pk. | Pack of 4 |
| 10424983 | 10 mm | 4 mm | 3 μm | Guard Cartridge, 4 Pk. | Pack of 4 |
| 10455733 | 10 mm | 2.1 mm | 3 μm | Guard Cartridge, 4 Pk. | Pack of 4 |
| 11969098 | 10 mm | 4 mm | 2.4 μm | Guard Cartridge, 4 Pk. | Pack of 4 |
| 11919098 | 10 mm | 2.1 mm | 2.4 μm | Guard Cartridge, 4 Pk. | Pack of 4 |
| 10203183 | 150 mm | 4.6 mm | 3 μm | Analytical Column | Each |
| 10027314 | 150 mm | 4 mm | 3 μm | Analytical Column | Each |
| 10558025 | 150 mm | 3 mm | 3 μm | Analytical Column | Each |
| 10099332 | 150 mm | 2.1 mm | 3 μm | Analytical Column | Each |
| 10374183 | 100 mm | 4.6 mm | 3 μm | Analytical Column | Each |
| 10723657 | 100 mm | 4 mm | 3 μm | Analytical Column | Each |
| 10666604 | 100 mm | 3 mm | 3 μm | Analytical Column | Each |
| 10243703 | 100 mm | 2.1 mm | 3 μm | Analytical Column | Each |
| 10563225 | 50 mm | 4.6 mm | 3 μm | Analytical Column | Each |
| 10478754 | 50 mm | 3 mm | 3 μm | Analytical Column | Each |
| 10495163 | 50 mm | 2.1 mm | 3 μm | Analytical Column | Each |
| 10511695 | 30 mm | 2.1 mm | 3 μm | Analytical Column | Each |
| 10269864 | 30 mm | 4.6 mm | 2.4 μm | Analytical Column | Each |
Description
For a robust, general-purpose column to use in a wide range of applications, choose Thermo Scientific™ Hypersil™ BDS C18 HPLC Columns. These economical columns feature reduced silanol activity and provide exceptional reliability for QA/QC labs. Built on the renowned Hypersil silica backbone, the base-deactivated silica is one of the most popular packing materials available.
For more challenging separations that require improved chromatographic performance, use Thermo Scientific™ Hypersil GOLD™ columns as an alternative.
- Original Hypersil columns from the manufacturer
- Base deactivated with minimal residual silanol activity
- Exceptional reproducibility
- Economical, general purpose columns
Ordering Information:
Other column dimensions are available. Please call Customer Service for more information.
Specifications
Specifications
| Column Format | Analytical Column |
| Diameter (Metric) | 2.1 mm |
| Length (Metric) | 50 mm |
| Max. Pressure | 5800 psi (400 bar) |
| Particle Size | 5 μm |
| pH | 2 to 8 |
| Pore Size | 130 Å |
| Surface Area | 170 m²/g |
| Temperature | 60°C |
| USP Type | L1 |
| Stationary Phase | C18 |
| Product Line | Hypersil BDS |
| Packing Material | Spherical, Fully Porous, Base-Deactivated Silica |
| Carbon Load | 11% |
| Column Type | Reversed Phase |
| Endcapped | Yes |
| Phase | Reversed Phase |
| Show More Show Less |