Learn More
Thermo Scientific™ HyperSep™ SLE Cartridges (pH 9)
Product Code. 12201178 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
1 g/6 mL, Pk. of 30, pH9
2 g/12 mL, Pk. of 20, pH9
20 g/60 mL, Pk. of 10, pH9
200 mg/3 mL, Pk. of 50, pH9
4 g/25 mL, Pk. of 15, pH9
500 mg/3 mL, Pk. of 50, pH9
500 mg/6 mL, Pk. of 30, pH9
Unit Size:
Pack of 10
Pack of 15
Pack of 20
Pack of 30
Pack of 50
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
12201178 500 mg/6 mL, Pk. of 30, pH9 Pack of 30
12201218 500 mg/3 mL, Pk. of 50, pH9 Pack of 50
12201198 200 mg/3 mL, Pk. of 50, pH9 Pack of 50
12201138 2 g/12 mL, Pk. of 20, pH9 Pack of 20
12201098 20 g/60 mL, Pk. of 10, pH9 Pack of 10
12201158 1 g/6 mL, Pk. of 30, pH9 Pack of 30
12201118 4 g/25 mL, Pk. of 15, pH9 Pack of 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 12201178 Supplier Thermo Scientific™ Supplier No. 6010950069
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
A fast and effective sample preparation technique for removal of phospholipids from biological matricies.
Provide a fast and effective sample preparation technique for removal of phospholipids from biological matrices with Thermo Scientific™ HyperSep™ SLE (Solid supported Liquid/Liquid Extraction).
HyperSep SLE cartrides (pH9) deals with sample preparation of biological matrices via a simple, efficient and an effective protcol. The focus being the removal of interfering phospholipids which cause a number of issues for end point analysis.
SLE offers a number of advantages over existing sample preparation techniques such as Liquid/Liquid Extraction (LLE) and Protein Precipitation techniques for dealing with biological matrices. In comparison to LLE, SLE offers greater reproducibility and recoveries, lower solvent requirements and prevents emulsification which is often associated with the LLE technique. It also lends itself to full automation which is not possible via LLE. SLE provides improved cleanliness of sample extract and associated increase in sensitivity compared to protein precipitation techniques which are unable to remove phospholipids.
  • Simple, easy-to-use protocol
  • Removal of phospholipids
  • Greater reproducability and recoveries compared to LLE
TRUSTED_SUSTAINABILITY
Specifications
Description 500 mg/6 mL, Pk. of 30, pH9
Bed Weight 500 mg
For Use With (Application) Removal of Phospholipids From Biological Matrices
Product Line HyperSep
Sorbent HyperSep Special Treated Diatomite
Type Solid Phase Extraction Cartridge
Volume (Metric) 6 mL
Quantity Pack of 30