Learn More
Thermo Scientific™ HyPerforma Tank Liners, CX3-9 Liner
Product Code. 17840567 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
19 L
28 L
38 L
378 L
757 L
Unit Size:
Pack of 10
Pack of 5
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
17840567 19 L Pack of 10
17810647 378 L Pack of 10
17820567 757 L Pack of 10
17830567 28 L Pack of 10
17870567 38 L Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17840567 Supplier Thermo Scientific™ Supplier No. SH30647.18
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
Tank liner system with top train is composed of CX3-9 single web, multi-layer plastic film, with dimensions optimized for drums and commonly used industry standard cylindrical tanks. They feature an open top for maximum recovery, with drum dolly available to provide mobility.
  • Minimizes bioburden due to construction in an ISO 7 cleanroom under sterile cGMP conditions.
  • Eliminates post-use cleaning steps required with re-useable containers and reduces cross-contamination risks.
Applications:
  • Removes the need for tank cleaning and helps reduce cycle times.
  • Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 19 L
Capacity (English) 5 gal.
Outer Packaging Cardboard Cartons
Protective Inner Packaging Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
Sterilization Method Irradiated
Product Line HyPerforma
Film Description CX3-9