Learn More
First Aid Kit Plasters
Product Code. 12950455
Change view
Click to view available options
Product Type:
HypaPlast Blue Catering Plaster
HypaPlast Pink Plaster
HypaPlast Plaster
Size:
7.2 x 2.5 cm
Assorted
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Size Unit Size
12950455 HypaPlast Plaster 7.2 x 2.5 cm Each
12930455 HypaPlast Plaster Assorted Each
12940455 HypaPlast Pink Plaster 7.2 x 2.5 cm Each
12920455 HypaPlast Pink Plaster Assorted Each
12960455 HypaPlast Blue Catering Plaster Assorted Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12950455 Supplier Safety First Supplier No. D8001
  • ONLINE EXCLUSIVE PRICE
    3.75 EUR
    2.65 EUR / Pack of 1
    Save 1.10 EUR
    29% Off
Estimated Shipment: 28-10-2026
Use First Aid Kit Plasters as an integral part of your first aid kit supplies for workplace, school, or home.
ul>
  • Various sizes, colors and styles
  • Included: Washproof plasters, fabric plasters and detectable blue plasters for catering industry
  • Strong, durable and highly conformable
  • Specifications
    Material Fabric
    Product Type HypaPlast Plaster
    Sterility Sterile
    Quantity 100 Pk.
    Size 7.2 x 2.5 cm
    Dimensions (L x W) 7.2 x 2.5 cm
    Packaging Type Individually Wrapped
    Waterproof Yes
    For Use With (Application) Multi-purpose, First aid kit