Learn More
Safety First Aid HypaClean Automatic Hand Gel Dispenser
Descrizione
- It is ideal for public places including businesses, hospitals, restaurants, stations and airports and due to the automatic feature it prevents individuals from touching the dispenser.
- Each output from the dispenser is approximately 0.5 mL to 0.6 mL if filled with hand gel.
- The delay between each output of hand gel is approximately 3 sec.
- Requires 4x C battery to allow the automatic feature to work, these are not included.
- The dispenser can be used approximately 12000 to 13000 times when running on a new set of batteries.
- Includes dispenser, key, screw and anchors, double sided tape and an instruction sheet.
- Dispenser supplied empty - gel must be purchased separately.
Specifica
Specifica
| Product Type | Hand Sanitizer Dispenser |
| Color | White |
| Holds | Hand Sanitizer |
| Material | Plastic |
| Mounting | Wall Mount |
| No. of Compartments | 1 |
| Product Style | Top Loading |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?