Learn More
Hyoscyamine HBr, MP Biomedicals
Product Code. 11427884 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11427884 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11427884 Supplier MP Biomedicals Supplier No. 0520772710
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
CAS 306-03-6
Molecular Formula C17H24BrNO3
Quantity 10 g
Synonym hyoscyamine hydrobromide, daturine hydrobromide, unii-iwt50p9s79, l-hyoscyamine hydrobromide, hyoscyamine hydrobromide usp, --atropine hydrobromide, c17h23no3.hbr, 1alphah,5alphah-tropan-3alpha-ol--tropate ester hydrobromide, unii-6vkw7r97zu component vzdnsfsbcmcxsk-pgqienjjsa-n
InChI Key VZDNSFSBCMCXSK-BXSBZORQSA-N
SMILES CN1C2CCC1CC(C2)OC(=O)C(CO)C3=CC=CC=C3.Br
IUPAC Name [(1S,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] (2S)-3-hydroxy-2-phenylpropanoate;hydrobromide
Molecular Weight (g/mol) 370.287
PubChem CID 9929278
Formula Weight 370.28
Chemical Name or Material Hyoscyamine HBr
Mehr anzeigen Weniger anzeigen