missing translation for 'onlineSavingsMsg'
Learn More

Hygromycin B Solution, ≥80%, MP Biomedicals™

Product Code. 11414022 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 mU
Unit Size:
5 micro units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11414022 5 mU 5 micro units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11414022 Supplier MP Biomedicals Supplier No. 0215751394
  • ONLINE EXCLUSIVE PRICE
    2230.00 EUR
    2121.00 EUR / 5 micro units
    Save 109.00 EUR
    4% Off
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An aminoglycoside antibiotic produced by the bacterium Streptomyces hygroscopicus that kills bacteria, fungi and higher eukaryotic cells by inhibiting protein synthesis

Specifications

CAS 31282-04-9
Color Yellow
Assay Percent Range ≥80%
Molecular Formula C20H37N3O13
MDL Number MFCD06795479
Quantity 5 mU
Synonym hygromycin b
InChI Key GRRNUXAQVGOGFE-BBMONYMYSA-N
SMILES CNC1CC(C(C(C1O)OC2C3C(C(C(O2)CO)O)OC4(O3)C(C(C(C(O4)C(CO)N)O)O)O)O)N
IUPAC Name (3'S,3aS,4S,4'S,5'R,6R,6'R,7R,7aS)-4-[(1R,2S,3R,5S,6S)-3-amino-2,6-dihydroxy-5-(methylamino)cyclohexyl]oxy-6'-[(1S)-1-amino-2-hydroxyethyl]-6-(hydroxymethyl)spiro[4,6,7,7a-tetrahydro-3aH-[1,3]dioxolo[4,5-c]pyran-2,2'-oxane]-3',4',5',7-tetrol
Molecular Weight (g/mol) 527.524
PubChem CID 134129613
Formula Weight 527.5
Physical Form Liquid
Chemical Name or Material Hygromycin B Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.