Learn More
Hygromycin B, 92.30%, For HPLC analysis, MP Biomedicals™
Brand: MP Biomedicals 0219417050_CLEAN
Safety and Handling
- Hygromycin B
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Respiratory sensitiser Category 1
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
Hazard Statement
- H300+H330-Fatal if swallowed or if inhaled
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P320-Specific treatment is urgent (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.