Learn More
Sampling Systems™ Hygienic Butterfly Valve Kit
Product Code. 17857817 Shop All Sampling Systems Products
Change view
Click to view available options
Includes:
Valve with EPDM Seals
Valve with Silicone Seals
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes unitSize
17857817 Valve with EPDM Seals Each
17912102 Valve with Silicone Seals Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17857817 Supplier Sampling Systems™ Supplier No. A13601A
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
These hygienic butterfly valves are manufactured from 316L stainless steel and are ideal for many applications, including GMP use.
  • Comes with a silicone or EPDM seal
  • Both seals are suitable for food and pharmaceutical use and come with relevant certification
  • Easy to operate: Simply turn the handle to open or close
  • Opening the handle slightly will mean you have reduced product flow
  • Valves are ideal for liquids
Specifications
Product Type Valve Kit
For Use With (Application) Dispensing Liquid
For Use With (Equipment) Drum with Sloping Base
Includes Valve with EPDM Seals