missing translation for 'onlineSavingsMsg'
Learn More

Hydroxyurea, (Crystalline),MP Biomedicals

Product Code. 11348849 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Unit Size:
10g
1g
25g
5g
Product Code. Quantity unitSize
11348849 1 g 1g
11358849 5 g 5g
11368849 10 g 10g
11378849 25 g 25g
4 options
This item is not returnable. View return policy

Product Code. 11348849

Brand: MP Biomedicals 0210202301

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibitor of DNA synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-07-1
Molecular Formula CH4N2O2
Molecular Weight (g/mol) 76.055
InChI Key VSNHCAURESNICA-UHFFFAOYSA-N
Synonym hydroxycarbamide, n-hydroxyurea, hydrea, 1-hydroxyurea, oxyurea, carbamoyl oxime, biosupressin, hydroxycarbamine, onco-carbide, carbamohydroxamic acid
PubChem CID 3657
ChEBI CHEBI:44423
IUPAC Name hydroxyurea
SMILES C(=O)(N)NO

Specifications

Quantity 1 g
Formula Weight 76.1
Physical Form Crystalline
Chemical Name or Material Hydroxyurea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.