Learn More
Hydroxyflutamide, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 52806-53-8 |
| Molecular Formula | C11H11F3N2O4 |
| MDL Number | MFCD00563126 |
| Quantity | 5 g |
| SMILES | CC(C)(O)C(NC1=CC=C([N+]([O-])=O)C(C(F)(F)F)=C1)=O |
| Molecular Weight (g/mol) | 292.21 |
| Percent Purity | 98% |
| Chemical Name or Material | Hydroxyflutamide |