missing translation for 'onlineSavingsMsg'
Learn More

Hydroxyapatite, TRC

Product Code. 30134339
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134339 25 g 25g
30137952 100 g 100g
30011976 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134339 Supplier Toronto Research Chemicals Supplier No. TRCH80190025G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12167-74-7
Molecular Formula 5 Ca . 3 O4 P . H O
Molecular Weight (g/mol) 502.31
Synonym Calcium hydroxide phosphate (Ca5(OH)(PO4)3), Calcium phosphate (Ca10(OH)2(PO4)6) (6CI), Calcium hydroxide phosphate (Ca10(OH)2(PO4)6), Calcium hydroxyphosphate (Ca10(OH)2(PO4)6), Calcium hydroxyphosphate (Ca5(PO4)3OH), Calcium phosphate (Ca5OH(PO4)3), Calcium phosphate hydroxide (Ca10(PO4)6(OH)2), Calcium phosphate hydroxide (Ca5(PO4)3(OH)), Calcium tribasic phosphate, Decacalcium hexaphosphate dihydroxide, Hydroxyapatite (synthetic), Pentacalcium hydroxide triphosphate, Pentacalcium hydroxylphosphate, Tribasic Calcium Phosphate
IUPAC Name pentacalcium;hydroxide;triphosphate
SMILES [OH-].[Ca+2].[Ca+2].[Ca+2].[Ca+2].[Ca+2].[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-]

Specifications

Quantity 25 g
InChI Formula InChI=1S/5Ca.3H3O4P.H2O/c;;;;;3*1-5(2,3)4;/h;;;;;3*(H3,1,2,3,4);1H2/q5*+2;;;;/p-10
Formula Weight 501.676
Chemical Name or Material Hydroxyapatite (synthetic)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.