missing translation for 'onlineSavingsMsg'
Learn More

Hydroxy Ibuprofen, TRC

Product Code. 30136713
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136713 100 mg 100mg
30040593 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136713 Supplier Toronto Research Chemicals Supplier No. TRCH943200100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53949-54-5
Molecular Formula C13 H18 O3
Molecular Weight (g/mol) 222.28
Synonym Benzeneacetic acid, 4-(3-hydroxy-2-methylpropyl)-α-methyl-, 2-[4-(3-Hydroxy-2-methyl-propyl)phenyl]propanoic acid, 4-(3-Hydroxy-2-methylpropyl)-α-methylbenzeneacetic acid, 2,4'-(2-Hydroxymethylpropyl)phenylpropionic acid, 2-[4-(2-Hydroxymethylpropyl)phenyl]propanoic acid, 3-Hydroxyibuprofen
IUPAC Name 2-[4-(3-hydroxy-2-methylpropyl)phenyl]propanoic acid
SMILES CC(CO)Cc1ccc(cc1)C(C)C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C13H18O3/c1-9(8-14)7-11-3-5-12(6-4-11)10(2)13(15)16/h3-6,9-10,14H,7-8H2,1-2H3,(H,15,16)
Formula Weight 222.1256
Chemical Name or Material 2-[4-(2-Hydroxymethylpropyl)phenyl]propanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.