Learn More
Hydroxocobalamin monohydrochloride, MedChemExpress
Product Code. 16434657
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16434657 50 mg 50mg
16418876 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16434657 Supplier Medchem Express Supplier No. HYB2209A
MedChemExpress Hydroxocobalamin monohydrochloride (Vitamin B12a monohydrochloride) is an injectable naturally occurring form of vitamin B12 with a favorable adverse effect profile, used as a dietary supplement in the treatment of vitamin B12 deficiency including pernicious anemia.
Hydroxocobalamin monohydrochloride (Vitamin B12a monohydrochloride) is an injectable naturally occurring form of vitamin B12 with a favorable adverse effect profile, used as a dietary supplement in the treatment of vitamin B12 deficiency including pernicious anemia.
Spezifikation
Chemical Name or Material Hydroxocobalamin monohydrochloride
CAS 59461-30-2
Color Black
Purity Grade Notes Research
Molecular Formula C62H90ClCoN13O15P
Quantity 50 mg
Synonym Vitamin B12a monohydrochloride
Solubility Information DMSO : 100 mg/mL (72.32 mM; Need ultrasonic) ∣H2O : 25 mg/mL (18.08 mM; Need ultrasonic)
SMILES CC12[N]([Co+3]34([N]5=CN(C(OC(CO)C6OP7([O-])=O)C6O)C8=C5C=C(C)C(C)=C8)([OH-])[N-]9C2C(CC(N)=O)C(C)(CCC(NCC(C)O7)=O)C9=C(C)C(C(CCC(N)=O)C%10(C)C)=[N]3C%10=CC%11=[N]4C(C(CC(N)=O)(C)C%11CCC(N)=O)=C%12C)=C%12C(CCC(N)=O)C1(C)CC(N)=O.Cl[H]
Molecular Weight (g/mol) 1382.82
Formula Weight 1382.82
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Mehr anzeigen Weniger anzeigen