Learn More
Hydrotalcite, synthetic, TRC
Product Code. 30137859
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137859 100 g 100g
30136483 50 g 50g
30012786 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137859 Supplier Toronto Research Chemicals Supplier No. TRCH714835100G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 11097-59-9
Molecular Formula CO3 . 6Mg . 4HO . 4H2O . 2AlH6O6
Molecular Weight (g/mol) 603.977
IUPAC Name hexamagnesium;hexahydroxyaluminum;carbonate;tetrahydroxide;tetrahydrate
SMILES O.O.O.O.[OH-].[OH-].[OH-].[OH-].[Mg+2].[Mg+2].[Mg+2].[Mg+2].[Mg+2].[Mg+2].O[Al](O)(O)(O)(O)O.O[Al](O)(O)(O)(O)O.[O-]C(=O)[O-]
Specifications
Quantity 100 g
Synonym Aluminate (Al(OH)63-), (OC-6-11)-, magnesium carbonate hydroxide (2:6:1:4), Magnesium, (carbonato)hexadecahydroxydialuminumhexa-, Aluminum, (carbonato)hexadecahydroxyhexamagnesiodi-, Carbonic acid, aluminum-magnesium complex, Magnesium aluminum hydroxycarbonate, Aluminum magnesium carbonate hydroxide, Magnesium aluminum hydroxy carbonate, Mg Al Hydrotalcite
InChI Formula InChI=1S/CH2O3.2Al.6Mg.20H2O/c2-1(3)4;;;;;;;;;;;;;;;;;;;;;;;;;;;;/h(H2,2,3,4);;;;;;;;;20*1H2/q;2*+6;6*+2;;;;;;;;;;;;;;;;;;;;/p-18
Formula Weight 601.9409
Chemical Name or Material Hydrotalcite, synthetic