Learn More
Hydroquinine, 95%
Product Code. 15763447 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15763447 1 g 1g
15773447 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15763447 Supplier Thermo Scientific Acros Supplier No. 461650010
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 522-66-7 | C20H26N2O2 | 326.44 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 522-66-7
Molecular Formula C20H26N2O2
Molecular Weight (g/mol) 326.44
MDL Number MFCD00151107
InChI Key LJOQGZACKSYWCH-ADMHDHIONA-N
Synonym dihydroquinine, hydroquinine, 1r-2s,4s,5r-5-ethylquinuclidin-2-yl 6-methoxyquinolin-4-yl methanol, unii-31j3q51t6l, hydrochinin, quinine, 10,11-dihydro, --dihydroquinine, --dihydroquinine, analytical standard, 8a,9r-10,11-dihydro-6'-methoxycinchonan-9-ol
PubChem CID 121515
IUPAC Name (R)-[(2S,4S,5R)-5-ethyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol
SMILES CCC1CN2CCC1CC2[C@H](O)C1=CC=NC2=CC=C(OC)C=C12
Specifications
Melting Point 168.0°C
Boiling Point 468.4°C
Flash Point 225°C
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 326.43
Percent Purity 95%
Chemical Name or Material Hydroquinine
compliance-icons
Product Identifier
  • Hydroquinine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Hydroquinine