accessibility menu, dialog, popup

UserName

CAS RN 522-66-7

NH3C(R)NOOHH3C

CAS RN 522-66-7

IUPAC Name: {5-ethyl-1-azabicyclo[2.2.2]octan-2-yl}(6-methoxyquinolin-4-yl)methanol
Synonyms: hydroquinidine
Molecular Weight (g/mol): 326.44
Molecular Formula: C20H26N2O2
InChi Key: LJOQGZACKSYWCH-UHFFFAOYNA-N
SMILES: CCC1CN2CCC1CC2C(O)C1=C2C=C(OC)C=CC2=NC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
Boiling Point 
  • (2)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results