Learn More
(-)-Dihydroquinine, TRC
Product Code. 30386591
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30386591 10 g
30381635 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30386591 Supplier Toronto Research Chemicals Supplier No. TRCD44991510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 522-66-7
Molecular Formula C20 H26 N2 O2
Molecular Weight (g/mol) 326.43
Synonym Cinchonan-9-ol, 10,11-dihydro-6'-methoxy-, (8α,9R)-, Hydroquinine (6CI,7CI,8CI), (-)-10,11-Dihydroquinine, (-)-Dihydroquinine, (-)-Hydroquinine, (8α,9R)-10,11-Dihydro-6'-methoxycinchonan-9-ol, 10,11-Dihydroquinine, Dihydroquinine, NSC 41799
IUPAC Name (R)-[(2S,4S,5R)-5-ethyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol
SMILES CC[C@H]1CN2CC[C@H]1C[C@H]2[C@H](O)c3ccnc4ccc(OC)cc34
Specifications
Quantity 10 g
InChI Formula InChI=1S/C20H26N2O2/c1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18/h4-6,8,11,13-14,19-20,23H,3,7,9-10,12H2,1-2H3/t13-,14-,19-,20+/m0/s1
Formula Weight 326.1994
Chemical Name or Material (-)-Dihydroquinine