Learn More
Hydrobromic Acid, 47.0%, Suprapur™, MilliporeSigma™
Product Code. 15816840
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15816840 1 L 1 L
15806840 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15816840 Supplier Supelco Supplier No. 1003061000
 Restricted Item
Estimated Shipment: 10-11-2026
Chemical Identifiers
CAS 10035-10-6
Molecular Formula BrH
Molecular Weight (g/mol) 80.91
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym Hydrogen bromide solution
IUPAC Name hydrogen bromide
SMILES Br
Specifications
Melting Point −11°C
Boiling Point 126°C (1013 hPa)
Density 1.49 g/cm3 (at 20°C)
Packaging Glass Bottle
Vapor Pressure 10.6 hPa (20°C)
Assay Percent Range ≥ 47%
Quantity 1 L