missing translation for 'onlineSavingsMsg'
Learn More

Hydrobenzamide, 98%

Product Code. 15410378
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15410378 25 g 25g
15400378 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15410378 Supplier Thermo Scientific Alfa Aesar Supplier No. H53492.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 92-29-5 | C21H18N2 | 298.389 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-29-5
Molecular Formula C21H18N2
Molecular Weight (g/mol) 298.389
MDL Number MFCD00022039
InChI Key VUYRFIIFTJICNA-LKNRODPVSA-N
Synonym hydrobenzamide, methanediamine, 1-phenyl-n,n'-bis phenylmethylene, n,.alpha.-diamine, toluene-.alpha.,.alpha.-diamine, n,n'-dibenzylidene, toluene-.alpha., n,n'-dibenzylidene, methanediamine,n'-bis phenylmethylene, n,n'-dibenzylidenetoluene-alpha,alpha-diamine, hydrobenzamid, n,n'-dibenzylidene-1-phenylmethanediamine
PubChem CID 9566477
IUPAC Name (E)-N-[[(E)-benzylideneamino]-phenylmethyl]-1-phenylmethanimine
SMILES C1=CC=C(C=C1)C=NC(C2=CC=CC=C2)N=CC3=CC=CC=C3

Specifications

Melting Point 102°C to 105°C
Quantity 25 g
UN Number UN2811
Beilstein 615231
Formula Weight 298.39
Percent Purity 98%
Chemical Name or Material Hydrobenzamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.