Learn More
HYDRANAL™-Sodium tartrate dihydrate, Solstice
Product Code. 15664430
Change view
Click to view available options
Quantity:
25 g
100 g
6 x 100 g
Unit Size:
100g
25g
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15664430 6 x 100 g Pack of 6
15644430 25 g 25g
15624430 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15664430 Supplier Honeywell-Hydranal Supplier No. 348036X100G
  • ONLINE EXCLUSIVE PRICE
    447.00 EUR
    416.97 EUR / Pack of 6
    Save 30.03 EUR
    6% Off
 Restricted Item
In Stock
Add to basket
Test substance for volumetric Karl Fischer titration (water content ∼15.66%, exact value on report of analysis)
Advantages of HYDRANAL- Auxiliaries
  • Broad product range of solubilizer for special sample application
  • Broad product range of buffer for pH related sample application
  • Drying agents for cell protection and drying of carrier gasses.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material HYDRANAL™-Sodium tartrate dihydrate
CAS 6106-24-7
Quantity 6 x 100 g
UN Number NONH for all modes of transport
Grade Karl Fischer