missing translation for 'onlineSavingsMsg'
Learn More

Hydramethylon, TRC

Product Code. 30135419
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135419 1 g 1g
30133951 10 g 10g
30012407 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135419 Supplier Toronto Research Chemicals Supplier No. TRCH6752001G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67485-29-4
Molecular Formula C25 H24 F6 N4
Molecular Weight (g/mol) 494.48
Synonym 2(1H)-Pyrimidinone, tetrahydro-5,5-dimethyl-, [3-[4-(trifluoromethyl)phenyl]-1-[2-[4-(trifluoromethyl)phenyl]ethenyl]-2-propenylidene]hydrazone (9CI), AC 217300, Amdro, Combat, Hydramethylnon, Maxforce, Maxforce Gel K, Maxforce Roach Killer Bait Gel, NSC 379665, 1,4-Pentadien-3-one, 1,5-bis[4-(trifluoromethyl)phenyl]-, 2-(1,4,5,6-tetrahydro-5,5-dimethyl-2-pyrimidinyl)hydrazone
IUPAC Name N-[[(1E,4E)-1,5-bis[4-(trifluoromethyl)phenyl]penta-1,4-dien-3-ylidene]amino]-5,5-dimethyl-1,3-diazinan-2-imine
SMILES CC1(C)CNC(=NN=C(\C=C\c2ccc(cc2)C(F)(F)F)\C=C\c3ccc(cc3)C(F)(F)F)NC1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C25H24F6N4/c1-23(2)15-32-22(33-16-23)35-34-21(13-7-17-3-9-19(10-4-17)24(26,27)28)14-8-18-5-11-20(12-6-18)25(29,30)31/h3-14H,15-16H2,1-2H3,(H2,32,33,35)/b13-7+,14-8+
Formula Weight 494.1905
Chemical Name or Material Hydramethylnon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.