Learn More
Cytiva HyClone™ SFM4MAb Powdered Media
Product Code. 10153252
Change view
Click to view available options
Quantity:
5 L
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10153252 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10153252 Supplier Cytiva Supplier No. SH30535.02
  • ONLINE EXCLUSIVE PRICE
    270.00 EUR
    233.53 EUR / 5 L
    Save 36.47 EUR
    13% Off
 Restricted Item
Estimated Shipment: 25-01-2027
Increase process yields for manufacturing of human and humanized recombinant antibodies.
SFM4MAb is a low protein formulation and is optimized for downstream purification using protein A, protein G, and other matrices to facilitate product recovery. The medium is formulated using proprietary lipid and phospholipid complexing process for enhanced stability and growth promotion of various cell types.
SFM4MAb is available in liquid and powder formats in user-friendly packaging.
  • Designed for high cell yield and antibody production
  • Complexed lipids for enhanced stability
  • Allows for direct or sequential adaptation
  • Optimized for protein A, protein G, and other purification matrices
  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
TRUSTED_SUSTAINABILITY
Specifications
Form Powder
Sterility Non-sterile
Quantity 5 L
Content And Storage 2 to 8°C, away from light and moisture