Learn More
Cytiva HyClone™ SFM4Insect Powdered Cell Culture Media
Product Code. 12954095
Change view
Click to view available options
Quantity:
5 L
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12954095 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12954095 Supplier Cytiva Supplier No. SH30912.01
  • ONLINE EXCLUSIVE PRICE
    321.00 EUR
    249.45 EUR / 5 L
    Save 71.55 EUR
    22% Off
 Restricted Item
Estimated Shipment: 02-02-2027
ADCF (animal-derived component-free) medium supports recombinant protein production with multiple insect cell lines.
SFM4Insect medium provides excellent growth of many key insect cell lines while requiring minimal adaptation. Additionally, SFM4Insect medium has been successfully tested in a variety of culture systems up to and including high density bioreactors.
  • For use with the baculovirus expression vector system (BEVS)
  • Designed for high cell yield and recombinant protein production
  • Complexed lipids for enhanced stability
  • Supports multiple insect cell lines
  • Supports direct or sequential adaptation
  • Manufactured using ISO 9001 and ISO 13485 certified processes according to 21 CFR 820 medical device current good manufacturing practices (cGMP)
TRUSTED_SUSTAINABILITY
Specifications
Form Powder
Sterility Non-sterile
With Additives Glutamine
Without Additives No Sodium Bicarbonate
Quantity 5 L
Content And Storage 2 to 8°C, away from light and moisture