Learn More
Cytiva HyClone™ SFM4CHO™ Media
Product Code. p-7058041
Click to view available options
Quantity:
500 mL
1000 mL
5 L
With Additives:
Glutamine, Sodium Bicarbonate, Poloxamer 188
Sodium Bicarbonate, Poloxamer 188
Without Additives:
No Glutamine, No Phenol Red, No HEPES
No Glutamine, No Sodium Bicarbonate, No Poloxamer 188, No Phenol Red, No HEPES
No Phenol Red, No HEPES
Form:
Liquid
Powder
Unit Size:
1000mL
5 L
500mL
Product Code. 10483522
Brand: Cytiva SH30549.02
Designed for high performance in a variety of culture vessels, including bioreactors. HyClone™ SFM4CHO™ Media is a protein-free formulation that contains no components of bovine origin.
  • Formulated using proprietary lipid complexing process for enhanced stability and growth promotion of various CHO cell lines
TRUSTED_SUSTAINABILITY
Specifications
Form Liquid
Product Type SFM4CHO Liquid Media
Sterilization Method 0.2 μm Sterile-filtered
With Additives Glutamine, Sodium Bicarbonate, Poloxamer 188
Without Additives No Phenol Red, No HEPES
Quantity 1000 mL
Certifications/Compliance cGMP
Content And Storage 2° to 8°C, away from light
Description SFM4CHO media